Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ret, Mouse anti-Human, Clone: RET/2663, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279761100UG
Description
Ret Monoclonal specifically detects Ret in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
Ret | |
Monoclonal | |
0.2 mg/mL | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array | |
P07949, P07949 | |
RET | |
Recombinant fragment (around aa 702-848) of human Ret protein (exact sequence is proprietary) | |
100 μg | |
Cancer, Oncogenes, Phospho Specific, Protein Kinase, Protein Phosphatase, Signal Transduction, Tumor Suppressors | |
5979 | |
Store at 4°C. | |
IgG2b κ |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
RET/2663 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
CDHF12MTC1, CDHR16, EC 2.7.10, Hirschsprung disease 1, Hirschsprung disease), MEN2A, MEN2B, multiple endocrine neoplasia and medullary thyroid carcinoma 1, ret proto-oncogene, RET transforming sequence | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title