Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ret, Mouse anti-Human, DyLight 350, Clone: RET/2663, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279896UV
Description
Ret Monoclonal antibody specifically detects Ret in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
Ret | |
RET/2663 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
RET | |
Recombinant fragment (around aa 702-848) of human Ret protein (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Store at 4°C in the dark. | |
Monoclonal |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CDHF12MTC1, CDHR16, EC 2.7.10, Hirschsprung disease 1, Hirschsprung disease), MEN2A, MEN2B, multiple endocrine neoplasia and medullary thyroid carcinoma 1, ret proto-oncogene, RET transforming sequence | |
Mouse | |
IgG2b Kappa | |
0.1 mL | |
Cancer, Oncogenes, Phospho Specific, Protein Kinase, Protein Phosphatase, Signal Transduction, Tumor Suppressors | |
Primary | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title