Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Retene, 97%, Acros Organics™

Click to view available options
Quantity:
250mg
1g
483-65-8
C18H18
234.34
NXLOLUFNDSBYTP-UHFFFAOYSA-N
retene,reten,1-methyl-7-isopropylphenanthrene,7-isopropyl-1-methylphenanthrene,phenanthrene, 1-methyl-7-1-methylethyl,1-methyl-7-1-methylethyl phenanthrene,unii-0w2d2e1p9q,phenanthrene, 7-isopropyl-1-methyl,ccris 3180,7-isopropyl-1-methylphenanthren
10222
1-methyl-7-propan-2-ylphenanthrene
CC1=CC=CC2=C1C=CC3=C2C=CC(=C3)C(C)C
483-65-8
C18H18
234.34
NXLOLUFNDSBYTP-UHFFFAOYSA-N
retene,reten,1-methyl-7-isopropylphenanthrene,7-isopropyl-1-methylphenanthrene,phenanthrene, 1-methyl-7-1-methylethyl,1-methyl-7-1-methylethyl phenanthrene,unii-0w2d2e1p9q,phenanthrene, 7-isopropyl-1-methyl,ccris 3180,7-isopropyl-1-methylphenanthren
10222
1-methyl-7-propan-2-ylphenanthrene
CC1=CC=CC2=C1C=CC3=C2C=CC(=C3)C(C)C
Melting Point 97.0°C to 99.0°C
Infrared Spectrum Authentic
Quantity 250mg
Boiling Point 108.0°C (10.0 mmHg)
Assay Percent Range 96% min. (HPLC)
Solubility Information Solubility in water: negligible. Other solubilities: soluble in hot alochol, benzene, hot ether, carbon, disulfide
Formula Weight 234.34
Percent Purity 97%
Chemical Name or Material Retene

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 207-597-9