Thermo Scientific™ Retention Gap Kits Available on GSA/VA Contract for Federal Government customers only.
Install a retention gap/guard column to your analytical column with a single kit.
Manufacturer: Thermo Scientific™ 260G500P
 View more versions of this product
Catalog No. 26-0G5-00P
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    For Use With GC
    Type GC Tubing
    Item Description Tubing; 0.53mm ID x 5m
    Description Tubing; 0.53mm ID x 5m
    Use Thermo Scientific™ Retention Gap Kits to conveniently install a retention gap/guard column to your analytical column. Each kit includes deactivated silica tubing, a miniunion, and five ferrules.
    Includes:
    • Length of deactivated fused silica tubing
    • Mini-union
    • Five ferrules
  • Description & Specifications
Related Products
Product Content Feedback