Learn More
Reticulon1A/B Mouse anti-Human, Janelia Fluor 646, Clone: RNL-3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP197735JF646
Catalog No. NBP97735646
Add to cart
Description
Description
Reticulon1A/B Monoclonal antibody specifically detects Reticulon1A/B in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
Specifications
Reticulon1A/B
RNL-3
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Purified
RTN1
RNL-3 is a Mouse monoclonal IgG1 antibody derived by fusion of SP2/0-Ag14 Mouse myeloma cells with spleen cells from a BALB/c Mouse immunized with the small cell lung cancer cell line NCI-H82. RNL-3 recognizes an epitope located within the region of amino
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
In normal tissues, RNL-3 reacts with brain Purkinje cells, pancreatic islet cells, cells in the pituitary gland and some (peripheral) nerve fibers. In addition, a few epithelia show positive staining.
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen)
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
reticulon-1, NSP, reticulon 1
Mouse
IgG1
0.1mL
Primary
6252
Human, Primate, Rabbit