Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RETN Rabbit anti-Human, Polyclonal, ProteinTech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 181701AP150UL
This item is not returnable.
View return policy
Description
Aliquoting is unnecessary for -20°C storage.
RETN Polyclonal antibody specifically detects RETN in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
RETN | |
Polyclonal | |
Unconjugated | |
RETN | |
Adipose tissue specific secretory factor; adipose tissue-specific secretory factor; Adipose-specific cysteine-rich secreted protein A12-alpha; ADSF; C/EBP epsilon regulated myeloid specific secreted cysteine rich protein precursor 1; C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich protein precursor 1; c/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein; cysteine-r; Cysteine-rich secreted protein A12-alpha-like 2; cysteine-rich secreted protein FIZZ3; dominant inhibitory adipocyte-specific secretory factor; dri; FIZZ3; found in inflammatory zone 3; HXCP1; MGC126603; MGC126609; Resistin; resistin delta2; RETN; RETN1; RSN; RSTN; UNQ407/PRO1199; XCP1; Xcp4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56729 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
250 μg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q9HD89 | |
RETN | |
RETN Fusion Protein Ag12822 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction