Learn More
Eisco™ Retort Stand Base and Rod Assembly
This premium steel “A” base provides a sturdy 3 point foundation for a wide array of lab activities that require stability and safety.
Specifications
Material Heavy duty cast steel
Thread Size 10 X 1.5mm
Color Blue
Shape “A”
View More Specs
Products 3
Catalog Number Mfr. No. Weight (Metric) Length (Metric) Base Length (Metric) Rod Price Quantity & Availability  
Catalog Number Mfr. No. Weight (Metric) Length (Metric) Base Length (Metric) Rod Price Quantity & Availability  
04-999-152 View Documents Eisco™
STDRDAB01/DEL
0.95kg 18cm 50cm This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
04-999-153 View Documents Eisco™
STDRDAB02/DEL
1.50kg 22cm 60cm This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
04-999-154 View Documents Eisco™
STDRDAB03/DEL
2.50kg 28cm 75cm This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Sturdy "A" shaped cast steel base that allows for unique arrangements and lab set up
  • Tapped hole receives a 10 X 1.5mm thread
  • Each side is finished in a chemical resistant coating to provide years of rugged laboratory use
Specifications
Specifications
Heavy duty cast steel
10 X 1.5mm
Blue
“A”