Tocris™ RETRA hydrochloride

Antitumor agent; suppresses mutant p53-bearing cancer cells

Manufacturer: Tocris™ 3214/10

 View more versions of this product

Catalog No. 32-141-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors RETRA hydrochloride
    Purity >99
    Molecular Weight 305.8
    Quantity 10mg
    Product Type RETRA hydrochloride

    Antitumor agent. Inhibits tumor cell growth in a mutant p53- and p73-dependent manner in vitro and in vivo (IC50 = 4µM). Transcriptionally activates expression of p53-responsive genes and induces activation of effector caspases 3 and 7.

    • Chemical Name: 2-[(4,5-Dihydro-2-thiazolyl)thio]-1-(3,4-dihydroxyphenyl)ethanone hydrochloride
    • Formula: C11N11NO3S2.HCl
    • CAS Number: 1173023-52-3
    • Solubility: Soluble to 100mM in DMSO
    • Purity: >99

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit