Learn More
3M™ Retrofit Pouch HF RFID Tag
3M™ Retrofit Pouch HF RFID Tag is best suited for PPE and other safety equipment that has structure, strap material or webbing of an acceptable width and thickness to accommodate the range of loop and snap size.
Manufacturer:  3M™ 7010315367
Catalog No. 19-071-128
Add to cart
Description
Description
  • Durable, heavy-duty construction
  • Pouch has a built-in choker and snap strap
  • RFID tag utilizes passive high frequency 13.56 MHz technology and is pre-programmed with a unique ID
  • Ready for use with 3M™ Safety and Inspection Manager
  • Includes an integral choker strap, snap strap, and zip ties
  • 3M Catalog# RFID-POUCH
Specifications
Specifications
Black
0.30 in
2.70 in
25/Pack
50.8 mm
Safety
Safety Program Management
0.8 cm
68.5 mm
2.0 in
Nylon
RFID Tags