Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Retrolinkin Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 151771AP

Catalog No. 50-173-0041


Add to cart

Description

Description

TMEM108 ft.s function has not been determined yet.
Specifications

Specifications

Retrolinkin
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6UXF1
KIAA1690, Retrolinkin, RTLN, TMEM108, transmembrane protein 108
Rabbit
IgG
150 μL
-20°C
Primary
66000
Western Blot
Unconjugated
TMEM108
Liquid
TMEM108
Retrolinkin Fusion Protein Ag7076
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit