Learn More
RETSCH PM 100 Planterary Ball Mill Grinding Jars
Specially designed for extreme working conditions such as long-term trials, wet grinding, high mechanical loads and maximum speeds as well as mechanical alloying
Manufacturer:  RETSCH 014620148
 View more versions of this product
Catalog No. 08-418-165
Add to cart
Description
Description
For use with Retsch PM 100 and PM 200 Planetary Ball Mills.
  • Safe, nonslip seating with built-in anti-rotation device and conical base centering
  • With O-ring for gastight and dustproof seal
  • Lid is easy to grip and open
Specifications
Specifications
4.22 oz.
4.22 oz., 125mL
Nonslip seating, built-in anti-rotation device, conical base centering, O-ring, dustproof seal
PM 100 and PM 200 planetary ball mill
125mL
Ideal for extreme working conditions such as long-term trials, wet grinding, high mechanical loads and maximum speeds as well as mechanical alloying
Stainless Steel
Grinding Jar