Learn More
RETSCH Test Sieves - 13 Mesh or Less
Stainless steel with wire gauze
Manufacturer:  RETSCH 60150063000
 View more versions of this product
Catalog No. 08-418-41
Add to cart
Description
Description
Dimensions: 8 dia. x 2 in.D (20.3 x 5cm)
Compliance
Meet ASTM E11
Specifications
Specifications
Ideal for cement clinker, chemicals, coffee, construction materials, fertilizers, fillers, flours, grains, Metals powders, Minerals, nuts, Plastics, sand, seeds, soils, washing powder etc.
5.08cm
63mm
ISO 3310
Stainless Steel
No. 2.5
2 in.
8 in.
Test Sieve
8 in.
2.5 Mesh
Stainless Steel