Learn More
RETSCH Test Sieves - 14 Mesh or Greater
Stainless steel with wire gauze
Supplier:  RETSCH 60150000045
 View more versions of this product
Catalog No. 08-417-95
Add to Cart
Description
Description
Specifications
Specifications
203 mm
Ideal for cement clinker, chemicals, coffee, construction materials, fertilizers, fillers, flours, grains, Metals powders, Minerals, nuts, Plastics, sand, seeds, soils, washing powder etc.
5.08cm
Test Sieve
8 in.
325 Mesh
Stainless Steel
2 in.
45μm
ASTM E11
Stainless Steel
No. 325
Product Suggestions
Product Suggestions