Learn More
RETSCH ZM 200 Mill Accessories
Manufacturer:  RETSCH 026080044
 View more versions of this product
Catalog No. 08-418-115
Add to cart
Description
Description
For use with ZM 200 Mixer Mill. Choose rotor based on properties of sample, required final fineness and subsequent analysis.
  • Stainless-steel, push-fit, with wear-resistant coating
  • 6-tooth rotor for coarse, bulky, fibrous goods such as feed pellets, hay and straw
  • 12-tooth rotor for medium-coarse goods such as wheat, oats, corn, tablets, powder coatings and plastics
Specifications
Specifications
Wear resistant coating
Stainless Steel
Ultra Centrifugal mill ZM 200
12 Teeth Rotor