Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RETSCH ZM 200 Ultra Centrifugal Mill, Push-Fit Rotors
For fine grinding of soft to medium-hard and fibrous materials.
$6530.00
Specifications
Material | Stainless Steel |
---|---|
Max. Speed | 6,000 to 18,000 rpm |
Max. Tube Diameter | 9.9 cm |
For Use With (Equipment) | Centrifugal Mill |
Description
- Accessory for use with ZM 200 Ultra Centrifugal Mill
- Wear resistant coating
Specifications
Stainless Steel | |
6,000 to 18,000 rpm | |
9.9 cm | |
Centrifugal Mill |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title