Learn More
SCS Reusable Caution Label - RS-471
Alerts personnel handling bags, boxes or other carriers that contents are static sensitive
Manufacturer:  SCS 7101
Catalog No. 18-688-268
Add to cart
Description
Description
  • Displays RS-471 ESD Susceptibility Symbol and warning messages
    Clearly identify products, packaging and areas requiring static protective measures
  • Reusable Label for reusable containers
  • Self-adhesive labels suitable for automatic dispensers and reel stock applications
    Label rolls include backing and paper core
  • Made in the United States of America
  • Recommended Storage Time: One year. Store at 72°F and 50% RH. Labels over one year old should be evaluated by the user to determine acceptability for the user’s application.
Specifications
Specifications
Not Autoclavable
Orange
ATTENTION STATIC-SENSITIVE DEVICES HANDLE ONLY AT STATIC-SAFE WORKSTATIONS REUSABLE CONTAINER DO NOT DESTROY
4.57 cm
6.35 cm
Paper
Non-sterile
Removable
JEDEC-14/EIA
Products, packages, and areas requiring static protective measures
Yes
1.8 x 2.5 in.
500/Roll