Learn More
Fisherbrand™ Reusable Glass Class A Volumetric Flasks with Snap Caps PROMO
Class A accuracy–without a serial number
 View more versions of this product
Catalog No. 10200CPM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Class A accuracy–without a serial number
  • Calibrated “To Contain”
Specifications
Specifications
Calibrated To Contain
Polyethylene
100 mL
Yes
±0.08 mL
Class A, ASTM E288
16
Glass
1