Learn More
Fisherbrand™ Reusable Glass Class A Volumetric Flasks with Standard Taper Stopper PROMO
Single graduation ring blasted on the neck
 View more versions of this product
Catalog No. 10205EPM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Single graduation ring blasted on the neck
  • Calibrated “To Contain”
  • Calibration indefinite if not exposed to hydrofluoric acid, hot phosphoric acid, or strong, hot alkalis
  • Do not heated above 150°C when dry
Specifications
Specifications
Calibrated To Contain
19
Glass
1
Glass
500 mL
Yes
±0.2 mL