Learn More
Fisherbrand™ Reusable Glass Narrow-Mouth Erlenmeyer Flasks PROMO
Long, tapered outside contour minimizes chipping
 View more versions of this product
Catalog No. FB500500PM
Add to cart
Description
Description
Fisherbrand™ Reusable Glass Narrow-Mouth Erlenmeyer Flasks are useful for measuring and mixing solutions where a high degree of accuracy is not necessary. Long, tapered outside contour minimizes chipping.
  • Reinforced top tooled with a rounded finish
  • Thick-walled body
  • White ceramic enamel marking spot
Specifications
Specifications
Rubber
Top style: Heavy duty tooled with capacity scale
102mm
Borosilicate Glass
100 to 500mL
6.85 in.
Narrow
Matte finish Ceramic Enamel marking
7
4.01 in.
500mL
No
50mL
17.4cm
1.0