Learn More
Fisherbrand™ Reusable Glass Narrow-Mouth Erlenmeyer Flasks PROMO
Long, tapered outside contour minimizes chipping
 View more versions of this product
Catalog No. FB50050PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
Description
Description
Fisherbrand™ Reusable Glass Narrow-Mouth Erlenmeyer Flasks are useful for measuring and mixing solutions where a high degree of accuracy is not necessary. Long, tapered outside contour minimizes chipping.
  • Reinforced top tooled with a rounded finish
  • Thick-walled body
  • White ceramic enamel marking spot
Specifications
Specifications
1
No
Top style: Heavy duty tooled with capacity scale
50 mL
20 to 50 mL
3.07 in.
1
50 mm
Rubber
1.96 in.
Borosilicate Glass
10 mL
7.8 cm
Matte Finish Ceramic Enamel Marking