Learn More
Fisherbrand™ Reusable Glass Narrow-Mouth Erlenmeyer Flasks PROMO
Long, tapered outside contour minimizes chipping
Specifications
Material Borosilicate Glass
No. of Positions 1
Closure Material Rubber
Disposable No
Mouth Style Narrow
View More Specs
Products 9
Catalog Number Capacity (Metric) Price Quantity & Availability  
Catalog Number Capacity (Metric) Price Quantity & Availability  
FB5001000PM View Documents 1000 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
FB500125PM View Documents 125 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
FB5002000PM View Documents 2000 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
FB50025PM View Documents 25 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
FB500250PM View Documents 250 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
FB5004000PM View Documents 4 L This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
FB50050PM View Documents 50 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
FB500500PM View Documents 500 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
FB5006000PM View Documents 6 L This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Fisherbrand™ Reusable Glass Narrow-Mouth Erlenmeyer Flasks are useful for measuring and mixing solutions where a high degree of accuracy is not necessary. Long, tapered outside contour minimizes chipping.
  • Reinforced top tooled with a rounded finish
  • Thick-walled body
  • White ceramic enamel marking spot
Specifications
Specifications
Borosilicate Glass
Rubber
Narrow
1
No
Matte Finish Ceramic Enamel Marking