Learn More
Fisherbrand™ Reusable Glass Wide-Mouth Erlenmeyer Flasks PROMO
High resistance to heat, chemical attack, and mechanical shock
Specifications
Material Borosilicate Glass
Disposable No
No. of Positions 1
Closure Material Rubber
Description Top style: Heavy duty tooled with capacity scale
View More Specs
Products 5
Catalog Number Capacity (Metric) Graduation Range Height (English) Closure Size Price Quantity & Availability  
Catalog Number Capacity (Metric) Graduation Range Height (English) Closure Size Price Quantity & Availability  
FB5011000PM View Documents 1000 mL 250 to 1000 mL 8.5 in. 11 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
FB501125PM View Documents 125 mL 50 to 125 mL 4.25 in. 6 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
FB5012000PM View Documents 2000 mL 600 to 2000 mL 10.4 in. 13 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
FB501250PM View Documents 250 mL 50 to 225 mL 5.11 in. 8 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
FB501500PM View Documents 500 mL 100 to 500 mL 6.77 in. 10 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Fisherbrand™ Reusable Glass Wide-Mouth Erlenmeyer Flasks offer sturdy borosilicate glass construction with a heavy-duty, tooled-top finish. High resistance to heat, chemical attack, and mechanical shock.
  • Capacity scale measures approximate volume
  • Matte-finish ceramic marking spot takes writing and erases easily
Specifications
Specifications
Borosilicate Glass
1
Top style: Heavy duty tooled with capacity scale
Wide
No
Rubber
Matte Finish Ceramic Enamel Marking