Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Black Stallion™ Tool Handz™ Plus Original Mechanics Gloves

Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves are constructed of lightweight and durable synthetic leather with a reinforced palm and fingertips.

  • Titan 2 Synthetic Leather Palm
  • Reinforced Contact Zones
  • Spandex Back
  • Strap Cuff
Back Material Spandex
Cuff Material Elastic
Coating Material Uncoated
Color Black
Hand Specific
Material Synthetic Leather
Product Type Mechanics Gloves
Size Small
For Use With (Application) Automotive Assembly, Construction, and Industrial Environments
Series 99PLUS