Learn More
Thermo Scientific™ Revco™ CxF Series -40°C Ultra-Low Temperature Chest Freezers
GSA_VA
Reliably store samples with versatile Thermo Scientific™ Revco™ CxF Series -40°C Ultra-Low Temperature Chest Freezers.
$9530.00
Specifications
Access Security Key Lock
Backup Systems CO2 or LN2 Optional
Certifications/Compliance CE, cULus, FDA Class I, UL
Chart Recorder Optional 7-day chart recorder
Control Type Push Button LED
View More Specs
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Products 3
Catalog Number Mfr. No. Holds Dimensions (DxWxH) Exterior Electrical Requirements Plug Type Price Quantity & Availability  
Catalog Number Mfr. No. Holds Dimensions (DxWxH) Exterior Electrical Requirements Plug Type Price Quantity & Availability  
ULT35010A
GSA_VA
Accessories View Documents
Thermo Scientific™
ULT35010A
Boxes (63) 29.9 x 28.5 x 43.8 in. (75.9 x 72.4 x 111.3 cm) 120 V 60 Hz NEMA 5-20
Each for $9,530.00
Add to cart
ULT135010A
Accessories View Documents
Thermo Scientific™
ULT135010A
Boxes (252) 33.4 x 72 x 40.5 in. (84.8 x 182.9 x 102.9 cm) 120 V 60 Hz NEMA 5-20 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ULT205010A
Accessories View Documents
Thermo Scientific™
ULT205010A
Boxes (396) 33.4 x 96 x 40.5 in. (84.8 x 243.8 x 102.9 cm) 120 V 60 Hz NEMA 5-20 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Reliably store samples with versatile Thermo Scientific™ Revco™ CxF Series -40°C ultra-low temperature chest freezers. Available in four convenient sizes and a full range of racking solutions, this unit delivers all the same sample protection features of our ExF and DxF freezers.
Features:
  • Easy-to-use microprocessor control panel
  • Access code to ensure setpoint security
  • Optional built-in chart recorder or data logger for continuous monitoring
  • Easy-to-remove air filter
  • Message center: 12-character alphanumeric LED displays setpoints, system status and alarm messages
Construction:
  • Heavy gauge, cold-rolled steel exterior with a powder coat paint finish that resists chipping and rust
  • 5 in. (127mm) foamed-in-place, polyurethane insulation
  • Stainless steel interior
  • Ergonomic, lockable door handle
Benefits:
  • Convenient interior covers minimize sample exposure to outside conditions.
  • Inner sub-lids protect bulk samples from ambient exposure during single sample retrievals.
  • Optional CO2 or LN2 safety back-up system is available for additional protection in the event of a power or mechanical failure.
  • RS-232 and 4-20 milliamp analog outputs are included for independent monitoring devices and recorders.
  • Freezer includes heavy-duty casters with locking tandem wheel design for smooth mobility
  • Warranty varies by country, please contact us for details
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Key Lock
CE, cULus, FDA Class I, UL
Push Button LED
Revco
Polyurethane Foam
Standard Operation
Smart-Vue Compatible
CO2 or LN2 Optional
Optional 7-day chart recorder
RS-485, 4-20mA, dry contacts - standard
-10°C to -40°C
Stainless Steel
Single ID
Buck/Boost