Thermo Scientific™ Revco™ Enzyme Freezer Available on GSA/VA Contract for Federal Government customers only.

Choose Thermo Scientific™ Revco™ Enzyme Freezers for storage applications where slight intermittent coil warming during an auto-defrost cycle is not desired.

Manufacturer: Thermo Scientific™ UEN2320A

 View more versions of this product

Catalog No. 13-990-206

  Check Availability
Add to Cart

    Specifications

    Warranty Warranty varies by country, please contact us for details
    Type Enzyme Freezer
    Doors Single
    Voltage 115V
    Width (English) Exterior 28 in.
    Width (English) Interior 24 in.
    Width (Metric) Exterior 71.1cm
    Width (Metric) Interior 61cm
    Certifications/Compliance Meets UL, CUL, and CE criteria for safety and performance
    Electrical Requirements 115V 60Hz
    Amperage 13.5A,13.5A
    Capacity 23.3 cu. ft., 659L,23.3 cu. ft., 659L
    Cabinet Material Scratch-resistant painted
    Breaker 20A,20A
    Capacity (English) 23.3 cu. ft.
    Capacity (Metric) 659L
    Chamber Material Scratch-resistant painted,Scratch-resistant painted
    Defrost Manual
    Depth (English) Interior 29 in.
    Depth (Metric) Interior 73.7cm
    Door Count 1,1
    Door Style Solid,Solid
    Dimensions (L x W x H) Exterior 37.2 x 28 x 79.2 in. (94.5 x 71.1 x 201.2cm)
    Dimensions (D x W x H) Interior 29 x 24 x 58 in. (73.7 x 61 x 147.3cm)
    Height (English) Interior 58 in.
    Height (English) Exterior 79.2 in.
    Height (Metric) Exterior 201.2cm
    Height (Metric) Interior 147.3cm
    Hertz 60Hz,60Hz
    Length (English) Exterior 37.2 in.,37.2 in.
    Operating Temperature -20°C,-20°C
    Length (Metric) Exterior 94.5cm,94.5cm
    Plug Type 5-20P,5-20P
    Shipping Weight (English) 425 lb.
    Shelves 9
    Refrigerant CFC-free,CFC-free
    Shipping Weight (Metric) 193kg
    Temperature Control Microprocessor

    Choose Thermo Scientific™ Revco™ Enzyme Freezers for storage applications where slight intermittent coil warming during an auto-defrost cycle is not desired. Our manual defrost -20°C upright freezers feature microprocessor control system, gravity air circulation, industrial-quality cabinet construction and extra-strength refrigeration compressors designed for industrial, clinical and scientific use.

    Key Features

    • Operating temperature: -20°C
    • Preset temperature setpoint
    • Upright 23.3 cu. ft. (659L) capacity
    • Gravity convection air system for maximum stability without blowers or fans
    • Manual defrost

    Microprocessor Control System:

    • Icon-based controls and displays
    • Graphic thermometer is steady green when temperature is normal, flashes top bar for overtemperature, flashes bulb for undertemperature
    • Large, easy-to-read digital display of temperature within 0.1°C
    • Audible/visual warnings for overtemperature, undertemperature, or power failure
    • Key-operated OFF/ON/ALARM ON master switch for setpoint security
    • Push-button alarm test
    • Remote alarm contacts
    • Alarm silence, ringback, and auto reset
    • Battery backup system

    Construction:

    • Scratch-resistant, painted cold-rolled steel interior and exterior finishes
    • Solid door with full-length handle and keyed lock—self-closes against closed-cell peripheral gasket
    • High-density, CFC-free urethane foam insulation protects cabinet from ambient temperature fluctuations and minimizes operating costs
    • Hermetically sealed heavy-duty, industrial-grade compressor
    • Nine epoxy-coated, open-wire shelves, adjustable in 1in. (2.5cm) increments
    • Adjustable leveling feet

    Ordering Information:

    Alarm systems, temperature chart recorders, and power monitors available separately. Contact your Customer Service or Sales Representative for details.

    Includes:

    45 high-impact plastic enzyme storage bins for easily accessible inventory. Bin overall dimensions (L x W x H): 25.25 x 4.25 x 3in. (64 x 11 x 8cm); labels not provided.

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit