Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Revco™ Enzyme Freezer Available on GSA/VA Contract for Federal Government customers only.

Choose Thermo Scientific™ Revco™ Enzyme Freezers for storage applications where slight intermittent coil warming during an auto-defrost cycle is not desired.

Manufacturer:  Thermo Scientific™ UEN2320D

 View more versions of this product

Catalog No. 13-990-654


  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Add to cart

Specifications

Description

Choose Thermo Scientific™ Revco™ Enzyme Freezers for storage applications where slight intermittent coil warming during an auto-defrost cycle is not desired. Our manual defrost -20°C upright freezers feature microprocessor control system, gravity air circulation, industrial-quality cabinet construction and extra-strength refrigeration compressors designed for industrial, clinical and scientific use.

Key Features
  • Operating temperature: -20°C
  • Preset temperature setpoint
  • Upright 23.3 cu. ft. (659L) capacity
  • Gravity convection air system for maximum stability without blowers or fans
  • Manual defrost

Microprocessor Control System:

  • Icon-based controls and displays
  • Graphic thermometer is steady green when temperature is normal, flashes top bar for overtemperature, flashes bulb for undertemperature
  • Large, easy-to-read digital display of temperature within 0.1°C
  • Audible/visual warnings for overtemperature, undertemperature, or power failure
  • Key-operated OFF/ON/ALARM ON master switch for setpoint security
  • Push-button alarm test
  • Remote alarm contacts
  • Alarm silence, ringback, and auto reset
  • Battery backup system

Construction:

  • Scratch-resistant, painted cold-rolled steel interior and exterior finishes
  • Solid door with full-length handle and keyed lock—self-closes against closed-cell peripheral gasket
  • High-density, CFC-free urethane foam insulation protects cabinet from ambient temperature fluctuations and minimizes operating costs
  • Hermetically sealed heavy-duty, industrial-grade compressor
  • Nine epoxy-coated, open-wire shelves, adjustable in 1in. (2.5cm) increments
  • Adjustable leveling feet

Ordering Information:

Alarm systems, temperature chart recorders, and power monitors available separately. Contact your Customer Service or Sales Representative for details.

Includes:

45 high-impact plastic enzyme storage bins for easily accessible inventory. Bin overall dimensions (L x W x H): 25.25 x 4.25 x 3in. (64 x 11 x 8cm); labels not provided.

Description & Specifications

Specifications

Meets UL, CUL, and CE criteria for safety and performance
23.3 cu. ft., 659L
15A
659L
Manual
73.7cm
Solid
29 x 24 x 58 in. (73.7 x 61 x 147.3cm)
79.2 in.
147.3cm
37.2 in.
94.5cm
425 lb.
CFC-free
Microprocessor
208 to 230V
24 in.
Single
208 to 230V 60Hz
5.6A
Scratch-resistant painted
23.3 cu. ft.
Scratch-resistant painted
29 in.
1
37.2 x 28 x 79.2 in. (94.5 x 71.1 x 201.2cm)
58 in.
201.2cm
60Hz
-20°C
6-15P
9
193kg
Enzyme Freezer
28 in.
71.1cm
61cm
Warranty varies by country, please contact us for details
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit