Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Revco™ Enzyme Freezer

Choose Thermo Scientific™ Revco™ Enzyme Freezers for storage applications where slight intermittent coil warming during an auto-defrost cycle is not desired.

Specifications

Cabinet Material Scratch-resistant painted
Defrost Manual
Depth (English) Interior 29 in.
Height (English) Exterior 79.2 in.
Height (English) Interior 58 in.
View More Specs

Products 2
Catalog Number Mfr. No. Capacity (English) Voltage Plug Type Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Voltage Plug Type Price Quantity & Availability  
UEN3020A
Accessories View Documents
Thermo Scientific™
UEN3020A
29.2 cu. ft. 115 V NEMA 5-15 P This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
13-990-206
Accessories View Documents
Thermo Scientific™
UEN2320A
23.3 cu. ft. 115 V NEMA 5-20 P This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Choose Thermo Scientific™ Revco™ Enzyme Freezers for storage applications where slight intermittent coil warming during an auto-defrost cycle is not desired. Our manual defrost -20°C upright freezers feature microprocessor control system, gravity air circulation, industrial-quality cabinet construction and extra-strength refrigeration compressors designed for industrial, clinical and scientific use.

Key Features
  • Operating temperature: -20°C
  • Preset temperature setpoint
  • Upright 23.3 cu. ft. (659L) capacity
  • Gravity convection air system for maximum stability without blowers or fans
  • Manual defrost

Microprocessor Control System:

  • Icon-based controls and displays
  • Graphic thermometer is steady green when temperature is normal, flashes top bar for overtemperature, flashes bulb for undertemperature
  • Large, easy-to-read digital display of temperature within 0.1°C
  • Audible/visual warnings for overtemperature, undertemperature, or power failure
  • Key-operated OFF/ON/ALARM ON master switch for setpoint security
  • Push-button alarm test
  • Remote alarm contacts
  • Alarm silence, ringback, and auto reset
  • Battery backup system

Construction:

  • Scratch-resistant, painted cold-rolled steel interior and exterior finishes
  • Solid door with full-length handle and keyed lock—self-closes against closed-cell peripheral gasket
  • High-density, CFC-free urethane foam insulation protects cabinet from ambient temperature fluctuations and minimizes operating costs
  • Hermetically sealed heavy-duty, industrial-grade compressor
  • Nine epoxy-coated, open-wire shelves, adjustable in 1in. (2.5cm) increments
  • Adjustable leveling feet

Ordering Information:

Alarm systems, temperature chart recorders, and power monitors available separately. Contact your Customer Service or Sales Representative for details.

Includes:

45 high-impact plastic enzyme storage bins for easily accessible inventory. Bin overall dimensions (L x W x H): 25.25 x 4.25 x 3in. (64 x 11 x 8cm); labels not provided.

  • Warranty varies by country, please contact us for details
  • Specifications

    Specifications

    Scratch-resistant painted
    29 in.
    58 in.
    Microprocessor
    37.2 in.
    -20°C
    Manual
    79.2 in.
    9
    Solid
    94.5 cm
    CFC-free
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit