Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Revco™ Flammable Storage Freezer, -20°C; 23 cu. ft., 115V

Protect flammable materials, solvents and intermediates using Revco™ FMS High-Performance Flammable Materials Freezers.

Manufacturer:  Thermo Scientific™ RFMS2320A

Catalog No. RFMS2320A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Description

Description

Protect flammable materials, solvents and intermediates using Thermo Scientific™ Revco FMS High-Performance Flammable Materials Freezers. These high-performance freezers are designed to meet stringent US Pharmacopeia standards for drug storage as well as the NFPA safety provisions for the storage of flammable liquids.† These freezers are designed to maintain a safe environment while keeping valuable samples within a tight temperature range. Each is listed by UL as a special purpose freezer intended for flammable material storage in accordance with NFPA 45, 70 and 99.† 45, 70 and 99.†

  • Easy to use intuitive digital display featuring microprocessor control
  • Lockable doors, audio and visual alarms including door ajar alarms for security and peace of mind
  • Convenient access port and casters
  • Outstanding construction featuring painted, high-quality steel exteriors with durable finish
  • Ignition-free interior cabinet design for the safe storage of flammable materials
  • Meets safety standards in accordance with UL, NFPA† and OSHA† for regulatory compliance
  • Optional, built-in, inkless, seven-day graphic chart recorder
  • Optional, barrier-protected 4-20 milliamp output for external monitoring
  • Manual defrost

Ordering Information

Temperature chart recorders, and 4-20 milliamp are available separately. Not Available Outside North America

Note:

† Products complying with the applicable requirements of NFPA 70, “National Electric Code”, NPFA 45, “Fire Protection for Laboratories Using Chemicals,” and NFPA 99 “Health Care Facilities” are considered to meet the OSHA guidelines for these types of facilities. These units have no internal electrical components that could trigger an explosion or fire of flammable materials inside the unit. Flammable material storage units are not for use in classified hazardous locations which require the use of explosion-proof units.

Specifications

Specifications

-20°C
23.3 cu. ft.
115 V 60 Hz
60 Hz
CFC-free
73.7 cm
58 in.
147.3 cm
61 cm
28 in.
37 in.
Lockable, Audio and Visual Alarms Including
CFC Free
External
4
CE, cUL, UL
659 L
115 V
29 x 24 x 58 in. (73.7 x 61 x 147.3 cm)
29 in.
78.6 in.
200 cm
24 in.
Manual
72 cm
94 cm
Scratch-resistant Painted Steel
Steel
1
SDS
Documents
Product Certifications
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit