Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Revco™ High-Performance Lab Freezers Available on GSA/VA Contract for Federal Government customers only.

Simplify operation and increase accuracy with the microprocessor control of Thermo Scientific™ Revco™ High-Performance Lab Freezers.

Manufacturer:  Thermo Scientific™ UGL2320A

 View more versions of this product

Catalog No. 13-990-205


  Check Availability
Add to cart

Specifications

Description

Simplify operation and increase accuracy with the microprocessor control of Thermo Scientific™ Revco™ High-Performance Lab Freezers. Our Revco™ -30°C and -20°C high-performance freezers are designed for medical and scientific applications such as storage of reagents, pharmaceuticals, biologicals or other commonly used laboratory materials. Models offer cabinet sizes to fit a variety of space needs and storage volume requirements.

  • Temperature factory-preset at -20° or -30°C
  • Capacities from 4.9 to 51.1 cu. ft. (139 to 1447L)
  • Icon-based controls and displays
  • Large, easy-to-read digital display shows temperature within 1°C
  • Graphic thermometer is steady green when temperature is normal, flashes top bar for overtemperature, flashes bulb for undertemperature

Microprocessor Control

  • Audible/visual warnings for overtemperature, undertemperature, or power failure
  • Key-operated OFF/ON/ALARM ON master switch for setpoint security
  • Push-button alarm test
  • Remote alarm contacts
  • Alarm silence, ringback, and auto reset
  • Controller battery backup system

Construction

  • Scratch-resistant cold-rolled steel interior and exterior finishes
  • High-density, 2in. thick (5.1cm) CFC-free urethane foam insulation protects against ambient temperature fluctuations
  • CFC-free, HFC-blend refrigerant
  • Single-door models are hinged on right
  • Full-length door handle and key lock
  • Doors self-close and lock open at 90°
  • Adjustable epoxy-coated wire shelves
  • 1in. (2.5cm) dia. access port with plug on undercounter model

Model Specific Features:

-30°C Freezers

  • Automatic defrost and condensate removal
  • ensor automatically initiates defrost cycle on demand, eliminating need for a defrost timer, minimizing frost buildup, and optimizing coil efficiency
  • Forced-air circulation improves temperature uniformity, provides faster recovery after door is opened

-20°C Freezers

  • Suitable for applications in which slight intermittent coil warming during an automatic defrost cycle is not desired (such as enzyme storage)
  • Gravity convection air system for maximum stability without a blower or fan
  • Manual defrost

Ordering Information:

Alarm systems, temperature chart recorders, and power monitors are available separately.

Note:

Published interior volumes are nominal; usable capacities may be offset by shelves, drawers, blower coils or cabinet protrusions. For specific cabinet dimensions and allowances, or for verification of clearance tolerances, contact your Customer Service Representative.

Description & Specifications

Specifications

UL and CUL listed. CE marked.
23.3 cu. ft., 659L
Upright cabinet
659L
29 in.
37.2 x 28 x 79.2 in. (94.5 x 71.1 x 201.2cm)
58 in.
201.2cm
60Hz
94.5cm
425 lb.
CFC-free
Microprocessor
High-Performance Freezer
28 in.
71.1cm
61cm
Warranty varies by country, please contact us for details
13.5A
20A
23.3 cu. ft.
Manual
73.7cm
29 x 24 x 58 in. (73.7 x 61 x 147.3cm)
79.2 in.
147.3cm
37.2 in.
5-20P
4
193kg
-20°C
115V
24 in.
Single, Solid
115V 60Hz
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit