Thermo Scientific™ Revco™ High-Performance Lab Freezers Available on GSA/VA Contract for Federal Government customers only.

Simplify operation and increase accuracy with the microprocessor control of Thermo Scientific™ Revco™ High-Performance Lab Freezers.

Manufacturer: Thermo Scientific™ ULT3030V

 View more versions of this product

Catalog No. 13-991-129

  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product.

Please contact customer service for assistance: 1-800-766-7000.

Add to Cart

    Specifications

    Warranty Warranty varies by country, please contact us for details
    Type High-Performance Freezer
    Voltage 220V
    Width (English) Interior 30 in.
    Width (Metric) Exterior 86.4cm
    Doors Single, Solid
    Width (Metric) Interior 76.2cm
    Amperage 12A,12A
    Capacity 29.2 cu. ft., 826L,29.2 cu. ft., 826L
    Capacity (English) 29.2 cu. ft.
    Capacity (Metric) 826L
    Depth (English) Interior 29 in.
    Dimensions (D x W x H) Interior 29 x 30 x 58 in. (73.7 x 76.2 x 147.3cm)
    Height (English) Interior 58 in.
    Height (English) Exterior 79.2 in.
    Height (Metric) Exterior 201.2cm
    Height (Metric) Interior 147.3cm
    Length (English) Exterior 37.2 in.
    Length (Metric) Exterior 94.5cm
    Plug Type 5-20P
    Shipping Weight (English) 445 lb.
    Shelves 4
    Shipping Weight (Metric) 202kg
    Temperature Control Microprocessor
    Width (English) Exterior 34 in.
    Certifications/Compliance UL and CUL listed. CE marked.
    Electrical Requirements 220V 50Hz
    Breaker 15A,15A
    Cabinet Type Upright cabinet,Upright cabinet
    Defrost Automatic
    Depth (Metric) Interior 73.7cm
    Dimensions (L x W x H) Exterior 37.2 x 34 x 79.2 in. (94.5 x 86.4 x 201.2cm)
    Hertz 50Hz
    Refrigerant CFC-free,CFC-free
    Temperature Range (Metric) -30°C

    Simplify operation and increase accuracy with the microprocessor control of Thermo Scientific™ Revco™ High-Performance Lab Freezers. Our Revco™ -30°C and -20°C high-performance freezers are designed for medical and scientific applications such as storage of reagents, pharmaceuticals, biologicals or other commonly used laboratory materials. Models offer cabinet sizes to fit a variety of space needs and storage volume requirements.

    • Temperature factory-preset at -20° or -30°C
    • Capacities from 4.9 to 51.1 cu. ft. (139 to 1447L)
    • Icon-based controls and displays
    • Large, easy-to-read digital display shows temperature within 1°C
    • Graphic thermometer is steady green when temperature is normal, flashes top bar for overtemperature, flashes bulb for undertemperature

    Microprocessor Control

    • Audible/visual warnings for overtemperature, undertemperature, or power failure
    • Key-operated OFF/ON/ALARM ON master switch for setpoint security
    • Push-button alarm test
    • Remote alarm contacts
    • Alarm silence, ringback, and auto reset
    • Controller battery backup system

    Construction

    • Scratch-resistant cold-rolled steel interior and exterior finishes
    • High-density, 2in. thick (5.1cm) CFC-free urethane foam insulation protects against ambient temperature fluctuations
    • CFC-free, HFC-blend refrigerant
    • Single-door models are hinged on right
    • Full-length door handle and key lock
    • Doors self-close and lock open at 90°
    • Adjustable epoxy-coated wire shelves
    • 1in. (2.5cm) dia. access port with plug on undercounter model

    Model Specific Features:

    -30°C Freezers

    • Automatic defrost and condensate removal
    • ensor automatically initiates defrost cycle on demand, eliminating need for a defrost timer, minimizing frost buildup, and optimizing coil efficiency
    • Forced-air circulation improves temperature uniformity, provides faster recovery after door is opened

    -20°C Freezers

    • Suitable for applications in which slight intermittent coil warming during an automatic defrost cycle is not desired (such as enzyme storage)
    • Gravity convection air system for maximum stability without a blower or fan
    • Manual defrost

    Ordering Information:

    Alarm systems, temperature chart recorders, and power monitors are available separately.

    Note:

    Published interior volumes are nominal; usable capacities may be offset by shelves, drawers, blower coils or cabinet protrusions. For specific cabinet dimensions and allowances, or for verification of clearance tolerances, contact your Customer Service Representative.

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit