Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Revco Industries™ Black Stallion™ 9 oz. FR Cotton Shop Coat

Manufacturer:  Revco Industries, IncSupplier Diversity Partner F942C

 View more versions of this product

Catalog No. 19-056558


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Snap
ASTM 6413
Medium
Cotton Fabric
106.6cm
2.0
122 to 127cm
0.2kg
Metal snap
Green
Reinforced snap front, inside pocket, scribe pocket and adjustable wrists
42 in.
Flame
48 to 50 in.
9 oz.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit