Learn More
Thermo Scientific™ Revco™ RDE Series Ultra-Low Temperature Freezers PROMO
Thermo Scientific™ Revco™ RDE Series ultra-low temperature freezers feature reduced energy usage, a simplified touchscreen interface, four capacities, and an ergonomic cabinet design.
$9200.00 - $9900.00
Specifications
Certifications/Compliance cULus
Defrost Manual
Temperature Range -50°C to -86°C
Product Line Revco
Style Upright
View More Specs
Products 4
Catalog Number Mfr. No. Capacity (Metric) Capacity (English) Electrical Requirements Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Capacity (English) Electrical Requirements Price Quantity & Availability  
RDE586FDQ4
View Documents
Thermo Scientific™
RDE50086FDQ4
682 L 24.1 cu. ft. 208 - 230 V 60 Hz
Each for $9,200.00
Add to cart
RDE586FAQ4
View Documents
Thermo Scientific™
RDE50086FAQ4
682 L 24.1 cu. ft. 115 V 60 Hz
Each for $9,200.00
Add to cart
RDE686FDQ4
View Documents
Thermo Scientific™
RDE60086FDQ4
816 L 28.8 cu. ft. 208 - 230 V 60 Hz
Each for $9,900.00
Add to cart
RDE686FAQ4
View Documents
Thermo Scientific™
RDE60086FAQ4
816 L 28.8 cu. ft. 115 V 60 Hz
Each for $9,900.00
Add to cart
Description
Description
Thermo Scientific™ Revco™ RDE Series ultra-low temperature freezers, powered by H-drive, feature four upright models, maximizing storage capacity from 30,000 up to 60,000 2mL vials. The RDE Series is a sustainable and reliable solution with ultimate sample security and operational savings for every lab.
Performance
  • Powered by H-drive's hydrocarbon technology, these systems save up to  25%  in energy usage and heat rejection over conventional refrigerant ultra-low freezers1
  • Designed for daily usage, recovers cabinet temperature 20% faster after opening the door.
  • A quiet sound output below 53 dBA, enables the freezer to come out of the hallway and into the lab
  • Meets +/-10°C validation protocols at -80°C with strong peak variation and steady-state temperature variation
  • Natural hydrocarbon refrigerants reduce system operating stresses promoting outstanding reliability and worry-free sample protection
User Friendly
  • The H-drive Information Center (HIC) combines easy to read temperature with simple capacitive touch buttons in a 5.6” display
  • Four capacities (300, 400, 500 and 600 2-inch box) to accommodate a variety of storage needs and lab footprint specifications
  • All models contain five inner doors allowing for easy sample access without exposing greater freezer content to ambient conditions
  • Improved customer convenience with new ergonomically designed handle, high contrast eye-level display, and comfortable easy-access shelf locations.
  • Compatible with most racking systems including a full range of Thermo Scientific™ racks for boxes, microplates, Thermo Scientific™ Matrix™ and Nunc CryoBank™ tubes
  • Security features including alarm settings and padlock-compatible doors
Sustainability
  • Manufactured in an award-winning, zero-waste to landfill facility
  • Lower HVAC costs as a result of less heat emission into the environment
  • Natural refrigerants and water-blown foam insulation, making the Series compliant with the U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP), the European Union’s F-Gas and other sustainability standards
Footnotes:
  1. Internal data comparing 11.2 kWh/day to UXF600 = 14.9 kWh/day. Data on file.
  2. Typical freezer test data based on internal testing in ambient temperature at 20°C; Typical standard validation protocol of +/-10°C.
Warranty and Services
Warranty varies by country, please contact us for details
Specifications
Specifications
cULus
-50°C to -86°C
Upright
Manual
Revco