Learn More
Thermo Scientific™ Revco™RLE ultra-low freezer with additional 3-year parts and labor warranty Available on GSA/VA Contract for Federal Government customers only.
Revco RLE ultra-low freezer package with additional 3-year parts and labor warranty (U.S. only)
Manufacturer:  Thermo Scientific™ RLE50086D5BB
 View more versions of this product
Catalog No. 09-313-840
Add to cart
Alert:
Not available outside North America
Description
Description
Revco RLE ultra-low freezer package with additional 3-year parts and labor warranty (U.S. only)
Package includes:
· One TSX 700 box capacity ultra-low freezer, 115V, 60Hz
· Fourteen sliding drawer racks with 280 2-inch cryoboxes
· Factory-installed chart recorder
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Warranty and Services
Warranty varies by country, please contact us for details
Specifications
Specifications
37.6 x 34 x 78 in.
Ultra Low Freezer, Upright
500 box capacity freezer with 3-year additional parts and labor warranty
−50° to −86°C
208-230V, 60Hz