Learn More
Thermo Scientific™ Revco™RLE ultra-low freezer with additional 3-year parts and labor warranty
Revco RLE ultra-low freezer package with additional 3-year parts and labor warranty (U.S. only)
Specifications
Temperature Range -50°C to -86°C
Type Ultra-Low Freezer, Upright
View More Specs
Alert:
Not available outside North America
Products 8
Catalog Number Mfr. No. Includes Electrical Requirements Type Price Quantity & Availability  
Catalog Number Mfr. No. Includes Electrical Requirements Type Price Quantity & Availability  
09-313-817 View Documents Thermo Scientific™
RLE30086A5BB
300 box capacity freezer with 3-year additional parts and labor warranty 115 V 60 Hz Ultra-Low Freezer, Upright This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
09-313-838 View Documents Thermo Scientific™
RLE30086D5BB
300 box capacity freezer with 3-year additional parts and labor warranty 208 - 230 V 60 Hz Ultra-Low Freezer, Upright This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
09-313-818 View Documents Thermo Scientific™
RLE40086A5BB
400 box capacity freezer with 3-year additional parts and labor warranty 115 V 60 Hz Ultra-Low Freezer, Upright This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
09-313-839 View Documents Thermo Scientific™
RLE40086D5BB
400 box capacity freezer with 3-year additional parts and labor warranty 208 - 230 V 60 Hz Ultra-Low Freezer, Upright This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
09-313-819 View Documents Thermo Scientific™
RLE50086A5BB
500 box capacity freezer with 3-year additional parts and labor warranty 115 V 60 Hz Ultra-Low Freezer, Upright This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
09-313-840 View Documents Thermo Scientific™
RLE50086D5BB
500 box capacity freezer with 3-year additional parts and labor warranty 208 - 230 V 60 Hz Ultra-Low Freezer, Upright This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
09-313-820 View Documents Thermo Scientific™
RLE60086A5BB
600 box capacity freezer with 3-year additional parts and labor warranty 115 V 60 Hz Ultra-Low Freezer, Upright This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
09-313-841 View Documents Thermo Scientific™
RLE60086D5BB
600 box capacity freezer with 3-year additional parts and labor warranty 208 - 230 V 60 Hz Ultra-Low Freezer, Upright This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Revco RLE ultra-low freezer package with additional 3-year parts and labor warranty (U.S. only)
Package includes:
  • One TSX 500 box capacity ultra-low freezer, 115V, 60Hz
  • Ten sliding drawer racks with 200 2-inch cryoboxes
  • Factory-installed chart recorder
Warranty and Services
Warranty varies by country, please contact us for details
Specifications
Specifications
-50°C to -86°C
Ultra-Low Freezer, Upright