Learn More
Thermo Scientific™ Revco™ RLE Ultra-Low Freezer Package with Racks and Boxes Available on GSA/VA Contract for Federal Government customers only.
Revco RLE ultra-low freezer package with racks and boxes
Manufacturer:  Thermo Scientific™ RLE40086ARAK
 View more versions of this product
Catalog No. 09-313-801
Add to cart
Description
Description
Revco™ RLE Ultra-Low temeprature freezer package with racks and boxes
Package includes:
· One Revco RLE 400 box capacity ultra-low freezer, 115V, 60Hz
· Eight sliding drawer racks with 160 2-inch cryoboxes
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Warranty and Services
Warranty varies by country, please contact us for details
Specifications
Specifications
−50° to −86°C
Ultra Low Freezer, Upright
115V, 60Hz
400 box capacity freezer, 8 sliding drawer racks, 200 2-inch boxes with 100-count cell dividers