Learn More
Thermo Scientific™ Revco™ RLE Ultra-Low temperature freezer package with racks, boxes, and HID access key lock
Revco RLE ultra-low freezer package including freezer, racks, boxes and factory-installed access key option
$19360.00 - $24770.00
Specifications
Temperature Range -50°C to -86°C
Type Ultra-Low Freezer, Upright
View More Specs
Alert:
Not available outside North America
Products 8
Catalog Number Mfr. No. Includes Electrical Requirements Type Price Quantity & Availability  
Catalog Number Mfr. No. Includes Electrical Requirements Type Price Quantity & Availability  
09-313-813 View Documents Thermo Scientific™
RLE30086ARAKHID
300 box capacity freezer, 6 sliding drawer racks, 150 2-inch boxes with 100-count cell dividers, and factory-installed HID access key 115 V 60 Hz Ultra-Low Freezer, Upright Each for $19,360.00
Add to cart
09-313-834 View Documents Thermo Scientific™
RLE30086DRAKHID
300 box capacity freezer, 6 sliding drawer racks, 150 2-inch boxes with 100-count cell dividers, and factory-installed HID access key 208 - 230 V 60 Hz Ultra-Low Freezer, Upright Each for $19,360.00
Add to cart
09-313-814 View Documents Thermo Scientific™
RLE40086ARAKHID
400 box capacity freezer, 8 sliding drawer racks, 200 2-inch boxes with 100-count cell dividers, and factory-installed HID access key 115 V 60 Hz Ultra-Low Freezer, Upright Each for $21,020.00
Add to cart
09-313-835 View Documents Thermo Scientific™
RLE40086DRAKHID
400 box capacity freezer, 8 sliding drawer racks, 200 2-inch boxes with 100-count cell dividers, and factory-installed HID access key 208 - 230 V 60 Hz Ultra-Low Freezer, Upright Each for $21,020.00
Add to cart
09-313-815 View Documents Thermo Scientific™
RLE50086ARAKHID
500 box capacity freezer, 10 sliding drawer racks, 250 2-inch boxes with 100-count cell dividers, and factory-installed HID access key 115 V 60 Hz Ultra-Low Freezer, Upright Each for $22,901.00
Add to cart
09-313-836 View Documents Thermo Scientific™
RLE50086DRAKHID
500 box capacity freezer, 10 sliding drawer racks, 250 2-inch boxes with 100-count cell dividers, and factory-installed HID access key 208 - 230 V 60 Hz Ultra-Low Freezer, Upright Each for $22,901.00
Add to cart
09-313-816 View Documents Thermo Scientific™
RLE60086ARAKHID
600 box capacity freezer, 12 sliding drawer racks, 300 2-inch boxes with 100-count cell dividers, and factory-installed HID access key 115 V 60 Hz Ultra-Low Freezer, Upright Each for $24,770.00
Add to cart
09-313-837 View Documents Thermo Scientific™
RLE60086DRAKHID
600 box capacity freezer, 12 sliding drawer racks, 300 2-inch boxes with 100-count cell dividers, and factory-installed HID access key 208 - 230 V 60 Hz Ultra-Low Freezer, Upright Each for $24,770.00
Add to cart
Description
Description
Revco™ RLE Ultra-Low temperature freezer package includes one ULT freezer, racks, boxes, and HID access key lock.
Package includes:
  • One Thermo Scientific Revco RLE Ultra-Low temperature freezer
  • 2 shelves of sliding drawer racks with 2-inch cryoboxes and 100-count cell dividers. (see product specifications for details)
  • Factory-installed HID access key lock system
Warranty and Services
Warranty varies by country, please contact us for details
Specifications
Specifications
-50°C to -86°C
Ultra-Low Freezer, Upright