Learn More
Thermo Scientific™ Revco™ RLE Ultra-Low temperature freezer package with racks, boxes, and chart recorder
Revco RLE ultra-low freezer package including freezer, racks, boxes and factory-installed chart recorder
$21987.00 - $25748.00
Specifications
Temperature Range -50°C to -86°C
Type Ultra-Low Freezer, Upright
View More Specs
Alert:
Not available outside North America
Products 6
Catalog Number Mfr. No. Includes Electrical Requirements Type Price Quantity & Availability  
Catalog Number Mfr. No. Includes Electrical Requirements Type Price Quantity & Availability  
09-313-810 View Documents Thermo Scientific™
RLE40086ARAKCR
400 box capacity freezer, 8 sliding drawer racks, 200 2-inch boxes with 100-count cell dividers, and factory-installed chart recorder 115 V 60 Hz Ultra-Low Freezer, Upright Each for $21,987.00
Add to cart
09-313-831 View Documents Thermo Scientific™
RLE40086DRAKCR
400 box capacity freezer, 8 sliding drawer racks, 200 2-inch boxes with 100-count cell dividers, and factory-installed chart recorder 208 - 230 V 60 Hz Ultra-Low Freezer, Upright Each for $21,987.00
Add to cart
09-313-811 View Documents Thermo Scientific™
RLE50086ARAKCR
500 box capacity freezer, 10 sliding drawer racks, 250 2-inch boxes with 100-count cell dividers, and factory-installed chart recorder 115 V 60 Hz Ultra-Low Freezer, Upright Each for $23,867.00
Add to cart
09-313-832 View Documents Thermo Scientific™
RLE50086DRAKCR
500 box capacity freezer, 10 sliding drawer racks, 250 2-inch boxes with 100-count cell dividers, and factory-installed chart recorder 208 - 230 V 60 Hz Ultra-Low Freezer, Upright Each for $23,867.00
Add to cart
09-313-812 View Documents Thermo Scientific™
RLE60086ARAKCR
600 box capacity freezer, 12 sliding drawer racks, 300 2-inch boxes with 100-count cell dividers, and factory-installed chart recorder 115 V 60 Hz Ultra-Low Freezer, Upright Each for $25,748.00
Add to cart
09-313-833 View Documents Thermo Scientific™
RLE60086DRAKCR
600 box capacity freezer, 12 sliding drawer racks, 300 2-inch boxes with 100-count cell dividers, and factory-installed chart recorder 208 - 230 V 60 Hz Ultra-Low Freezer, Upright Each for $25,748.00
Add to cart
Description
Description
Revco™ RLE Ultra-Low temperature freezer package includes one ULT freezer, racks, boxes, and a factory-installed chart recorder.
Package includes:
  • One Thermo Scientific Revco RLE Ultra-Low temperature freezer
  • 2 shelves of sliding drawer racks with 2-inch cryoboxes and 100-count cell dividers. (see product specifications for details)
  • Factory-installed chart recorder
Warranty and Services
Warranty varies by country, please contact us for details
Specifications
Specifications
-50°C to -86°C
Ultra-Low Freezer, Upright