Learn More
Thermo Scientific™ Revco™ RLE Ultra-Low temperature freezer package with racks, boxes, and LN2 back-up system
Revco RLE ultra-low freezer package including freezer, racks, boxes and factory-installed LN2 back-up
$21250.00 - $26736.00
Specifications
Temperature Range -50°C to -86°C
Type Ultra-Low Freezer, Upright
View More Specs
Alert:
Not available outside North America
Products 6
Catalog Number Mfr. No. Includes Electrical Requirements Type Price Quantity & Availability  
Catalog Number Mfr. No. Includes Electrical Requirements Type Price Quantity & Availability  
09-313-804 View Documents Thermo Scientific
RLE40086ARAKLN
400 box capacity freezer, 8 sliding drawer racks, 200 2-inch boxes with 100-count cell dividers, and factory-installed LN2 back-up system 115 V 60 Hz Ultra-Low Freezer, Upright Each for $21,250.00
Add to cart
09-313-825 View Documents Thermo Scientific™
RLE40086DRAKLN
400 box capacity freezer, 8 sliding drawer racks, 200 2-inch boxes with 100-count cell dividers, and factory-installed LN2 back-up system 208 - 230 V 60 Hz Ultra-Low Freezer, Upright Each for $22,984.00
Add to cart
09-313-805 View Documents Thermo Scientific
RLE50086ARAKLN
500 box capacity freezer, 10 sliding drawer racks, 250 2-inch boxes with 100-count cell dividers, and factory-installed LN2 back-up system 115 V 60 Hz Ultra-Low Freezer, Upright Each for $24,855.00
Add to cart
09-313-826 View Documents Thermo Scientific™
RLE50086DRAKLN
500 box capacity freezer, 10 sliding drawer racks, 250 2-inch boxes with 100-count cell dividers, and factory-installed LN2 back-up system 208 - 230 V 60 Hz Ultra-Low Freezer, Upright Each for $24,855.00
Add to cart
09-313-806 View Documents Thermo Scientific™
RLE60086ARAKLN
600 box capacity freezer, 12 sliding drawer racks, 300 2-inch boxes with 100-count cell dividers, and factory-installed LN2 back-up system 115 V 60 Hz Ultra-Low Freezer, Upright Each for $26,736.00
Add to cart
09-313-827 View Documents Thermo Scientific™
RLE60086DRAKLN
600 box capacity freezer, 12 sliding drawer racks, 300 2-inch boxes with 100-count cell dividers, and factory-installed LN2 back-up system 208 - 230 V 60 Hz Ultra-Low Freezer, Upright Each for $26,736.00
Add to cart
Description
Description
Revco™ RLE Ultra-Low temperature freezer package includes one ULT freezer, racks, boxes, and a factory-installed LN2 back-up system.
Package includes:
  • One Thermo Scientific Revco RLE Ultra-Low temperature freezer
  • 2 shelves of sliding drawer racks with 2-inch cryoboxes and 100-count cell dividers. (see product specifications for details)
  • Factory-installed LN2 back-up system (LN2 tank not included)
Warranty and Services
Warranty varies by country, please contact us for details
Specifications
Specifications
-50°C to -86°C
Ultra-Low Freezer, Upright