Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Black Stallion™ Select Shoulder Split Cowhide Leather Welder's Gloves with Cotton Lining
Click to view available options
Size:
Small
Description
- Select shoulder split cowhide leather construction is reliably consistent
- Cotton sock lining for additional comfort and thermal protection
- Aramid Fiber stitching for spark and abrasion resistance
- Thumb strap reinforcement provides extra durability
- Ideal protection for standard stick welding applications
Specifications
Color | Orange |
Material | Shoulder Split Cowhide Leather |
Size | Small |
For Use With (Application) | Stick Welding |
Glove Cuff Style | Safety |
Liner Material | Cotton |
Series | 110 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction