Learn More
BUCHI FlashPure ID Amino Flash Cartridges Available on GSA/VA Contract for Federal Government customers only.
Better peak shapes with polar and basic compounds, ideal for carbohydrates
Manufacturer:  BUCHI 145157331
 View more versions of this product
Catalog No. 05-404-822
Add to cart
Includes: 1 Piece
Description
Description
High performance
  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
Lower purification costs
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
  • /ul>
    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
12 g
1 Pc.
200 psig
Better Peak Shapes with Polar and Basic Compounds, Ideal for Carbohydrates
Silica - Amine
Reveleris