Learn More
BUCHI FlashPure ID Amino Flash Cartridges Available on GSA/VA Contract for Federal Government customers only.
Better peak shapes with polar and basic compounds, ideal for carbohydrates
$95.00 - $146.00
Specifications
Material Silica - Amine
Max. Pressure 200 psig
Type Reveleris
View More Specs
Includes: 1 Piece
Products 2
Catalog Number Mfr. No. Bed Weight (Metric) Includes Price Quantity & Availability  
Catalog Number Mfr. No. Bed Weight (Metric) Includes Price Quantity & Availability  
05-404-821 Available on GSA/VA Contract for Federal Government customers only.
View Documents
BUCHI
145157330
4 g 1 Pc.
Each for $95.00
Add to cart
05-404-822 Available on GSA/VA Contract for Federal Government customers only.
View Documents
BUCHI
145157331
12 g 1 Pc.
Each for $146.00
Add to cart
Description
Description
High performance
  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
Lower purification costs
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
  • /ul>
    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Silica - Amine
200 psig
Reveleris