Learn More
BUCHI FlashPure ID C18-WP Flash Chromatography Cartridges
FlashPure ID cartridges are designed for BUCHI Pure chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system.
Supplier: BUCHI 145169482
Includes: 1 Piece
Description
- High resolution of separation due to high-quality silica
- Increased fraction purity due to a better separation
- Faster evaporation of smaller fraction volumes
- Reproducible performance from batch to batch
- Increased sample throughput due to high sample loading capacity
- Solvent saving: required purification yield is achieved faster
Specifications
Silica - C-18 WP | |
1 Pc. | |
Ideal for Peptides, Proteins and Lipids and Other Large Molecule Purifications |
Reveleris | |
4 g | |
200 psig |