Learn More
BUCHI FlashPure ID C18-WP Flash Chromatography Cartridges
SDP
FlashPure ID cartridges are designed for BUCHI Pure chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system.
Supplier:  BUCHI 145169483
 View more versions of this product
Catalog No. 05-404-840
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: 1 Piece
Description
Description
  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
Specifications
Specifications
Silica - C-18 WP
1 Pc.
Ideal for Peptides, Proteins and Lipids and Other Large Molecule Purifications
Reveleris
12 g
200 psig