Learn More
BUCHI FlashPure ID C18-WP Flash Cartridges
Ideal for peptides, proteins and lipids
Manufacturer:  BUCHI 145169484
 View more versions of this product
Catalog No. 05-404-841
Includes: 1 Piece
Description
Description
High performance
  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
Lower purification costs
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
  • /ul>
Specifications
Specifications
Silica - C-18 WP
1 Pc.
Ideal for Peptides, Proteins and Lipids and Other Large Molecule Purifications
Reveleris
40 g
200 psig