Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BUCHI FlashPure ID C18-WP Flash Chromatography Cartridges

FlashPure ID cartridges are designed for BUCHI Pure chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system.
Specifications
Material | Silica - C-18 WP |
---|---|
Type | Reveleris |
Includes | 1 Pc. |
For Use With (Application) | Ideal for Peptides, Proteins and Lipids and Other Large Molecule Purifications |
Max. Pressure | 200 psig |
Includes: 1 Piece
Catalog Number | Mfr. No. | Bed Weight (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Bed Weight (Metric) | Price | Quantity & Availability | |||||
05-404-839
![]() |
BUCHI
145169482 |
4 g | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
05-404-840
![]() |
BUCHI
145169483 |
12 g | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- High resolution of separation due to high-quality silica
- Increased fraction purity due to a better separation
- Faster evaporation of smaller fraction volumes
- Reproducible performance from batch to batch
- Increased sample throughput due to high sample loading capacity
- Solvent saving: required purification yield is achieved faster
Specifications
Silica - C-18 WP | |
1 Pc. | |
200 psig |
Reveleris | |
Ideal for Peptides, Proteins and Lipids and Other Large Molecule Purifications |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title