Learn More
BUCHI FlashPure ID Silica Flash Chromatography Cartridges
SDP
FlashPure ID cartridges are designed for BUCHI Pure chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system.
Supplier:  BUCHI 145165750
 View more versions of this product
Catalog No. 05-404-838
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Includes: 20 Pieces
This item is not returnable. View return policy
Description
Description
  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
Specifications
Specifications
Silica
6 Pc.
Useful for Conventional Applications; Polar Compounds
Reveleris
220 g
100 psig