Learn More
BUCHI FlashPure ID HP Silica Flash Chromatography Cartridges
SDP
FlashPure ID cartridges are designed for BUCHI Pure chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system.
Manufacturer:  BUCHI 145170200
 View more versions of this product
Catalog No. 05-404-842
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: 20 Pieces
Description
Description
  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
Specifications
Specifications
Silica HP
20 Pc.
Superior Resolution for Complex or Difficult to Purify Samples
FlashPure
4 g
200 psig