Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI FlashPure ID HP Silica Flash Chromatography Cartridges
SDP

FlashPure ID cartridges are designed for BUCHI Pure chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system.

Manufacturer:  BUCHI 145170207

 View more versions of this product

Catalog No. 05-405-581

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

High Performance

  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch

Lower Purification Costs

  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
Specifications

Specifications

Silica HP
6 Pc.
220 g
Pure Chromatography Systems
FlashPure
High, Irregular (16 to 24 μm)
Superior Resolution for Complex or Difficult to Purify Samples
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit