Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI FlashPure ID HP Silica Flash Chromatography Cartridges Available on GSA/VA Contract for Federal Government customers only.

FlashPure ID cartridges are designed for BUCHI Pure chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system.

$251.00 - $728.50

Specifications

For Use With (Application) Superior Resolution for Complex or Difficult to Purify Samples
Material Silica HP
Max. Pressure 200 psig
Type FlashPure
View More Specs

Products 7
Catalog Number Mfr. No. Bed Weight (Metric) Includes Price Quantity & Availability  
Catalog Number Mfr. No. Bed Weight (Metric) Includes Price Quantity & Availability  
05-404-842
View Documents
BUCHI
145170200
4 g 20 Pc.
Each for $251.00
Add to cart
 
05-404-843 Available on GSA/VA Contract for Federal Government customers only.
View Documents
BUCHI
145170201
12 g 20 Pc.
Each for $351.00
Add to cart
 
05-404-844
View Documents
BUCHI
145170202
24 g 15 Pc.
Each for $384.00
Add to cart
 
05-404-845
View Documents
BUCHI
145170203
40 g 15 Pc.
Each for $523.00
Add to cart
 
05-404-846
View Documents
BUCHI
145170204
80 g 12 Pc.
Each for $631.00
Add to cart
 
05-404-847
View Documents
BUCHI
145170205
120 g 10 Pc.
Each for $728.50
Add to cart
 
05-405-581
View Documents
BUCHI
145170207
220 g 6 Pc. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
Specifications

Specifications

Superior Resolution for Complex or Difficult to Purify Samples
Silica HP
200 psig
FlashPure
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit