Learn More
Sklar InstrumentsSupplier Diversity Partner Reverdin Needle, Sklar, Slightly Curved, 9 in., Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 261951
Reverdin Needle, Sklar, Slightly Curved, 9 in., Stainless Steel. Sklar's Reverdin Needle is an instrument that resembles a needle holder and needle combination that acts as a suture passer. The eye of the Reverdin needle can be adjusted by a sliding mechanism on the handle, thus making it easier to deliver suture material. This assists the user by shortening the difficulty of suturing as well as the time necessary to complete suturing. This product features a slight curve and overall length of 9 inches.
Catalog No. 50-143-8073
Due to product restrictions, please to purchase or view availability for this product.